Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

493716

Sigma-Aldrich

Bis(cyclopentadienyl)manganese(II)

97%

Synonym(s):

Manganocene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Mn(C5H5)2
CAS Number:
Molecular Weight:
185.12
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

assay

97%

reaction suitability

core: manganese

mp

175 °C (lit.)

SMILES string

[Mn].[CH]1[CH][CH][CH][CH]1.[CH]2[CH][CH][CH][CH]2

InChI

1S/2C5H5.Mn/c2*1-2-4-5-3-1;/h2*1-5H;

InChI key

QROCSKKOEBOLQO-UHFFFAOYSA-N

comparable product

Product No.
Description
Pricing

pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Flam. Sol. 1 - Skin Irrit. 2 - STOT SE 3

Storage Class

4.1B - Flammable solid hazardous materials

wgk_germany

WGK 3

flash_point_f

125.6 °F - closed cup

flash_point_c

52 °C - closed cup

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service