Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

433233

Sigma-Aldrich

Nitric acid

70%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
HNO3
CAS Number:
Molecular Weight:
63.01
MDL number:
PubChem Substance ID:
Pricing and availability is not currently available.

agency

meets analytical specifications of BP
meets analytical specifications of E339
meets analytical specifications of Ph. Eur.
meets analytical specifications of Ph. Franç.
meets analytical specifications of USP

reg. compliance

meets analytical specifications of FCC

vapor pressure

8 mmHg ( 20 °C)

assay

70%

availability

available only in EU

bp

120.5 °C (lit.)

density

1.413 g/mL at 20 °C (lit.)

SMILES string

O[N+]([O-])=O

InChI

1S/HNO3/c2-1(3)4/h(H,2,3,4)

InChI key

GRYLNZFGIOXLOG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Eye Dam. 1 - Met. Corr. 1 - Ox. Liq. 3 - Skin Corr. 1A

supp_hazards

Storage Class

5.1B - Oxidizing hazardous materials

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wen Song et al.
Lab on a chip, 15(16), 3314-3325 (2015-07-08)
Sandstone formations are ubiquitous in both aquifers and petroleum reservoirs, of which clay is a major constituent. The release of clay particles from pore surfaces as a result of reduced injection fluid salinity can greatly modify the recovery of hydrocarbons
Souleymane Coulibaly et al.
Biological & pharmaceutical bulletin, 38(9), 1395-1403 (2015-09-04)
To clarify the seasonal fluctuations in air pollution and the effect of long-range transport, we collected airborne particles (n=118) at Dazaifu in Fukuoka, Japan, from June 2012 to May 2013 and measured Pb and SO4(2-), which are indicators of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service