Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

391204

Sigma-Aldrich

Methyl pipecolinate hydrochloride

97%

Synonym(s):

Methyl 2-piperidinecarboxylate hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H13NO2 · HCl
CAS Number:
Molecular Weight:
179.64
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

97%

mp

205 °C (dec.) (lit.)

functional group

ester

SMILES string

Cl.COC(=O)C1CCCCN1

InChI

1S/C7H13NO2.ClH/c1-10-7(9)6-4-2-3-5-8-6;/h6,8H,2-5H2,1H3;1H

InChI key

APCHKWZTSCBBJX-UHFFFAOYSA-N

General description

Methyl pipecolinate hydrochloride is a hydrochloride salt of methyl piperidine-2-carboxylate (methyl pipecolinate). The kinetics of the enzymatic separation of enantiomeric forms of methyl pipecolinate using Candida antarctica Lipase A (CAL-A) has been reported.[1][2] Its role as catalyst for the standard Diels-Alder reaction has been examined.[3]

Application

Reactant for synthesis of:
A pipecolic linker
Antiviral agents
Aurora and epidermal growth factor receptor kinase inhibitor
Pyrrolidine derivatives via reduction of substituted pyrroles

Reactant for:
Petasis reactions
Decarbonylative radical cyclization of alpha-amino selenoesters upon electrophilic alkenes

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The α-effect in cyclic secondary amines: new scaffolds for iminium ion accelerated transformations.
Brazier JB, et al.
Tetrahedron, 65(48), 9961-9966 (2009)
Enantioselective lipase-catalyzed reactions of methyl pipecolinate: transesterification and N-acylation.
Liljeblad, A, et al.
Tetrahedron Letters, 43(13), 2471-2474 (2002)
Alkoxycarbonylpiperidines as N-nucleophiles in the palladium-catalyzed aminocarbonylation.
Takacs A, et al.
Monatshefte fur Chemie / Chemical Monthly, 145(9), 1473-1478 (2014)
Aldehyde-based racemization in the dynamic kinetic resolution of N-heterocyclic a-amino esters using Candida Antarctica lipase A.
Liljeblad A, et al.
Tetrahedron, 60(3), 671-677 (2004)
Advances in the kinetic and dynamic kinetic resolution of piperazine-2-carboxylic acid derivatives with Candida antarctica lipase A; structural requirements for enantioselective N-acylation.
Hietanen A, et al.
ARKIVOC (Gainesville, FL, United States), 5, 60-74 (2012)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service