Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

390313

Sigma-Aldrich

Octafluoro-2-butanone

≥97%

Synonym(s):

Perfluoro-2-butanone

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$520.00

$520.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$520.00

About This Item

Linear Formula:
CF3CF2COCF3
CAS Number:
Molecular Weight:
216.03
Beilstein/REAXYS Number:
1790282
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$520.00


Check Cart for Availability

Request a Bulk Order

Quality Level

assay

≥97%

mp

−126 °C (lit.)

functional group

fluoro
ketone

SMILES string

FC(F)(F)C(=O)C(F)(F)C(F)(F)F

InChI

1S/C4F8O/c5-2(6,4(10,11)12)1(13)3(7,8)9

InChI key

QJPLLYVRTUXAHZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant involved in synthesis of 2,3-bis(perfluoroalkyl)quinoxalines

Other Notes

For 390313-100G-EU packaging: 100 mL of aluminium cylinder equipped with brass valve.

Recommended products

For 390313-100G-EU: outlet fitting is DIN 477-1 no.1, Z742160 or Z742126 are the recommended regulators.

pictograms

Gas cylinderSkull and crossbones

signalword

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Eye Irrit. 2 - Press. Gas Compr. Gas - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

2A - Gases

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service