Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

364916

Sigma-Aldrich

5-Fluoro-2-methylbenzonitrile

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FC6H3(CH3)CN
CAS Number:
Molecular Weight:
135.14
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

98%

form

solid

mp

43-45 °C (lit.)

functional group

fluoro
nitrile

SMILES string

Cc1ccc(F)cc1C#N

InChI

1S/C8H6FN/c1-6-2-3-8(9)4-7(6)5-10/h2-4H,1H3

InChI key

IBRODYNXELBTJC-UHFFFAOYSA-N

General description

5-Fluoro-2-methylbenzonitrile is an aryl fluorinated building block.

pictograms

CorrosionExclamation mark

signalword

Danger

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

174.2 °F - closed cup

flash_point_c

79 °C - closed cup

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service