Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

295965

Sigma-Aldrich

(Trimethylsilyl)acetonitrile

99%

Synonym(s):

(Cyanomethyl)trimethylsilane, TMSAN

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3SiCH2CN
CAS Number:
Molecular Weight:
113.23
Beilstein/REAXYS Number:
1738845
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

99%

refractive index

n20/D 1.420 (lit.)

bp

65-70 °C/20 mmHg (lit.)

density

0.827 g/mL at 25 °C (lit.)

SMILES string

C[Si](C)(C)CC#N

InChI

1S/C5H11NSi/c1-7(2,3)5-4-6/h5H2,1-3H3

InChI key

WJKMUGYQMMXILX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

(Trimethylsilyl)acetonitrile has been used in catalytic cyanomethylation of various carbonyl compounds in the presence of Lewis bases[1].

pictograms

FlameSkull and crossbones

signalword

Danger

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

109.4 °F - closed cup

flash_point_c

43 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Lewis base-catalyzed cyanomethylation of carbonyl compounds with (trimethylsilyl) acetonitrile.
Kawano Y, et al.
Chemistry Letters (Jpn), 34(11), 1508-1509 (2005)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service