Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

242330

Sigma-Aldrich

Dipentyl ether

97%

Synonym(s):

Amyl ether, Pentyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3(CH2)4]2O
CAS Number:
Molecular Weight:
158.28
Beilstein/REAXYS Number:
1698030
EC Number:
MDL number:
UNSPSC Code:
12190000

assay

97%

refractive index

n20/D 1.412 (lit.)

bp

187-188 °C (lit.)

mp

−69 °C (lit.)

density

0.785 g/mL at 25 °C (lit.)

SMILES string

CCCCCOCCCCC

InChI

1S/C10H22O/c1-3-5-7-9-11-10-8-6-4-2/h3-10H2,1-2H3

InChI key

AOPDRZXCEAKHHW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service