Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

42570

Sigma-Aldrich

Dipentyl ether

≥98.5% (GC)

Synonym(s):

Amyl ether, Pentyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3(CH2)4]2O
CAS Number:
Molecular Weight:
158.28
Beilstein/REAXYS Number:
1698030
EC Number:
MDL number:
UNSPSC Code:
12352112
PubChem Substance ID:
assay:
≥98.5% (GC)
technique(s):
GC/GC: suitable
bp:
187-188 °C (lit.)

assay

≥98.5% (GC)

contains

~0.0005% 2,6-di-tert-butyl-4-methylphenol as stabilizer

expl. lim.

2.0-8.0 %

technique(s)

GC/GC: suitable

refractive index

n20/D 1.412 (lit.)
n20/D 1.412

bp

187-188 °C (lit.)

mp

−69 °C (lit.)

density

0.785 g/mL at 25 °C (lit.)

SMILES string

CCCCCOCCCCC

InChI

1S/C10H22O/c1-3-5-7-9-11-10-8-6-4-2/h3-10H2,1-2H3

InChI key

AOPDRZXCEAKHHW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dipentyl ether (1,1-Dipentyl ether, DPeE) is an aliphatic diether. It is formed as one of the major product during the dehydration of pentan-1-ol.[1]

Application

Dipentyl ether has been used to compose the binary mixtures with various imidazole derivatives. Solid-liquid equilibrium (SLE) of these binary mixtures have been evaluated by a dynamic method.[2]

pictograms

Flame

signalword

Warning

hcodes

Hazard Classifications

Flam. Liq. 3

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

134.6 °F - closed cup

flash_point_c

57 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Solubility of imidazoles in ethers.
Domanska U and Kozlowska MK.
Journal of Chemical and Engineering Data, 48(3), 557-563 (2003)
Volumetric properties of (N,N-dimethylformamide + aliphatic diethers) at temperatures ranging from (298.15 to 358.15) K.
Venkatesu P, et al.
The Journal of Chemical Thermodynamics, 37(9), 996-1002 (2005)
Characterization and catalytic properties of saponite clay modified by acid activation.
Kooli F and Jones W.
Clay Minerals, 32(4), 633-643 (1997)
Andrzej Lech Dawidowicz et al.
Journal of separation science, 40(14), 2984-2991 (2017-05-31)
The quantitative relationship between analytes established by the headspace solid-phase microextraction procedure for multicomponent mixtures depends not only on the character and strength of interactions of individual components with solid-phase microextraction fiber but also on their vapor pressure in the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service