Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

220132

Sigma-Aldrich

Glyoxal trimeric dihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H6O6 · 2H2O
CAS Number:
Molecular Weight:
210.14
Beilstein/REAXYS Number:
3542160
EC Number:
MDL number:
UNSPSC Code:
12352100

SMILES string

O.O.OC1OC2OC(O)C(O)OC2OC1O


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service