151688
L-Histidine
98%
Synonym(s):
(S)-2-Amino-3-(4-imidazolyl)propionic acid, NSC 137773
About This Item
Recommended Products
assay
98%
optical activity
[α]23/D −38.4°, c = 3 in H2O
mp
282 °C (dec.) (lit.)
SMILES string
N[C@@H](Cc1c[nH]cn1)C(O)=O
InChI
1S/C6H9N3O2/c7-5(6(10)11)1-4-2-8-3-9-4/h2-3,5H,1,7H2,(H,8,9)(H,10,11)/t5-/m0/s1
InChI key
HNDVDQJCIGZPNO-YFKPBYRVSA-N
Looking for similar products? Visit Product Comparison Guide
Biochem/physiol Actions
Application
replaced by
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Chromatograms
application for HPLCOur team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service