Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

137855

Sigma-Aldrich

1,3-Dimethyl-5-nitrobenzene

≥99%

Synonym(s):

5-Nitro-m-xylene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C6H3NO2
CAS Number:
Molecular Weight:
151.16
Beilstein/REAXYS Number:
1936130
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

≥99%
≥99%

form

solid

bp

273 °C/739 mmHg (lit.)

mp

72-74 °C (lit.)

SMILES string

Cc1cc(C)cc(c1)[N+]([O-])=O

InChI

1S/C8H9NO2/c1-6-3-7(2)5-8(4-6)9(10)11/h3-5H,1-2H3

InChI key

BYFNZOKBMZKTSC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1,3-Dimethyl-5-nitrobenzene was used in aerobic oxidation of aromatic anilines to aromatic azo compounds using gold nanoparticles supported on TiO2 as a catalyst[1].

pictograms

Skull and crossbonesHealth hazard

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - STOT RE 2

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P2 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Abdessamad Grirrane et al.
Nature protocols, 5(3), 429-438 (2010-03-06)
This protocol describes the aerobic oxidation of aromatic anilines to aromatic azo compounds using gold (Au) nanoparticles supported on TiO(2) as a catalyst. Yields above 98% are achieved under a few bars of oxygen pressure. It should be noted that

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service