Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

376930

Sigma-Aldrich

2,6-Diisopropylphenyl isocyanate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)2CH]2C6H3NCO
CAS Number:
Molecular Weight:
203.28
Beilstein/REAXYS Number:
2645721
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

97%

form

liquid

refractive index

n20/D 1.519 (lit.)

bp

117 °C/10 mmHg (lit.)

density

0.951 g/mL at 25 °C (lit.)

functional group

isocyanate

SMILES string

CC(C)c1cccc(C(C)C)c1N=C=O

InChI

1S/C13H17NO/c1-9(2)11-6-5-7-12(10(3)4)13(11)14-8-15/h5-7,9-10H,1-4H3

InChI key

FEUFNKALUGDEMQ-UHFFFAOYSA-N

Gene Information

human ... GABRA1(2554)

Looking for similar products? Visit Product Comparison Guide

General description

Reaction of the complex trans-(PMe3)2Pd(Ph)(NHPh) with 2,6-diisopropylphenyl isocyanate has been reported.[1]

Application

2,6-Diisopropylphenyl isocyanate may be used in the preparation of:
  • mono- and dinuclear methylrhenium(VII) imido complexes[2]
  • [2]rotaxane[3]
  • 2,6-diisopropylphenyl-carbodiimide[4]

signalword

Danger

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Resp. Sens. 1 - Skin Corr. 1B - STOT SE 3

target_organs

Respiratory system

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 2

flash_point_f

224.6 °F - closed cup

flash_point_c

107 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Multiple bonds between transition metals and main-group elements. 72. Organorhenium imido complexes: syntheses, structure, and reactivity.
Herrmann WA, et al.
Organometallics, 9(2), 489-496 (1990)
Reactions of unsaturated electrophiles with trans-(PMe3)2 Pd (Ph)(NHPh).
Boncella JM and Villanueva LA.
Journal of Organometallic Chemistry, 465(1), 297-304 (1994)
Synthesis of 2, 6-Diisopropylphenyl-Carbodiimide.
Ye D-Y and Dai J-Y.
He Cheng Hua Xue / Chinese Journal of Synthetic Chemistry, 14(2), 200-200 (2006)
A new protocol for rotaxane synthesis.
Cantrill SJ, et al.
Tetrahedron Letters, 40(19), 3669-3672 (1999)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service