58453
1 of 10
Supelco
5.33002
甲酸98%-100%
Millipore
SLFGX13
Millex™疏水PTFE针式过滤器
5.43804
甲酸98-100%
55023
Aura 流动相过滤器/脱气器装置
64835-U
标准微型起泡器
58062-U
Supelco ® 流动相过滤装置 2
58061
Supelco® 流动相过滤装置 1
23207
节省空间型存放架
58093
24758
用于卡口样品瓶的瓶盖
universal design with bracket
從最近期的版本中選擇一個:
It looks like we've run into a problem, but you can still download Certificates of Analysis from our 文件 section.