重要文件
22517
Supelco
备用毛细管柱螺母
pkg of 2 ea
登入查看組織和合約定價
About This Item
分類程式碼代碼:
41121815
暫時無法取得訂價和供貨情況
包裝
pkg of 2 ea
一般說明
These brass adapters enable you to use long or general purpose ferrules in Agilent MSDs. Made of brass to prevent damage to MSD source threads. Knurled head allows tightening with fingers, eliminating the need to locate the proper wrench.
從最近期的版本中選擇一個:
分析證明 (COA)
Lot/Batch Number
如果您需要協助,請聯絡 客戶支援
已經擁有該產品?
您可以在文件庫中找到最近購買的產品相關文件。
存取文件庫
Debora Keller et al.
Nanoscale, 12(44), 22511-22517 (2020-11-12)
Elementary atomic mechanisms underlying nanoparticle growth in liquids are largely unexplored and mostly a subject of conjectures based on theory and indirect experimental insights. Direct, experimental observation of such processes at an atomic level requires imaging with single-atom sensitivity and