重要文件
Z741693
Sartorius low-retention pipette tips
volume range 50-1,000 μL, rack
登入查看組織和合約定價
About This Item
分類程式碼代碼:
41121600
NACRES:
NB.01
暫時無法取得訂價和供貨情況
材料
colorless
無菌
sterile
特點
DNase free
RNase free
barrier
endotoxin free
low retention
包裝
pack of 960 ea
製造商/商標名
Sartorius LH-LF791001
最大體積
1 mL
容積範圍
50-1,000 μL
結合類型
(liquid-repellent surface)
尋找類似的產品? 前往 產品比較指南
一般說明
Sartorius Low Retention Tips have an extremely even and durable liquid-repellent surface for optimising the flow of the liquid while dispensing, for optimum sample recovery. Liquids with low surface tension, like detergents, slide easily off of the inner surface of the tip, leaving very little residue behind. This Low Retention feature is thus very useful in the following applications, where such viscous, detergent containing, liquids are often pipetted: PCR, real-time PCR, cloning, SDS-PAGE and other nucleic acid or protein analysis methods. The Ttps are designed for best possible fitting and sealing to secure the highest possible accuracy and precision. These pipette tips enable ergonomic and light tip attachment by being compatible with the Optiload feature in Sartorius Picus, eLINE®, mLINE® and Proline® Plus pipettes. Tip trays are colour-coded for easy matching with corresponding, colour-coded Sartorius pipettes.
其他說明
Browse all pipette tips
法律資訊
Proline is a registered trademark of Sartorius Biohit Liquid Handling Oy
eLINE is a registered trademark of Sartorius Biohit Liquid Handling Oy
mLINE is a registered trademark of Sartorius Biohit Liquid Handling Oy
從最近期的版本中選擇一個:
分析證明 (COA)