重要文件
Z741669
Sartorius VALUEPAK pipette tips
volume range 0.1-10 μL
登入查看組織和合約定價
About This Item
分類程式碼代碼:
41121600
NACRES:
NB.01
暫時無法取得訂價和供貨情況
材料
colorless
無菌
non-sterile
特點
DNase free
RNase free
barrier: no
endotoxin free
包裝
pack of 960 ea (Refil)
pkg of (refill Tower)
製造商/商標名
Sartorius 790012MAX
最大體積
10 μL
容積範圍
0.1-10 μL
容積
0.1—10 μL
尋找類似的產品? 前往 產品比較指南
一般說明
Large value packs of tips with the perfect tip fitting and sealing to the pipetter. This gives you the best possible accuracy and precision at the best possible price. Compatible with Optiload feature in Biohit Picus, eLINE®, mLINE® and Proline® Plus pipetters, enabling ergonomic and light tip attachment and ejection.
其他說明
Browse all pipette tips
法律資訊
Proline is a registered trademark of Sartorius Biohit Liquid Handling Oy
eLINE is a registered trademark of Sartorius Biohit Liquid Handling Oy
mLINE is a registered trademark of Sartorius Biohit Liquid Handling Oy
從最近期的版本中選擇一個:
分析證明 (COA)