重要文件
Z741653
Sartorius filter pipette tips
volume range 0.1-10 μL, rack of, sterile
登入查看組織和合約定價
About This Item
分類程式碼代碼:
41121601
NACRES:
NB.01
暫時無法取得訂價和供貨情況
材料
colorless
polypropylene (Virgin)
無菌
sterile
特點
DNase free
RNase free
barrier
endotoxin free
filter
包裝
pack of 960 ea (Racked)
rack of
製造商/商標名
Sartorius 783201
最大體積
10 μL
容積範圍
0.1-10 μL
結合類型
low binding surface
尋找類似的產品? 前往 產品比較指南
一般說明
The Sartorius filter tips of the Biohitfamily are made of polypropylene and feature a polyethylene filter barrier to prevent aerosol and liquid contamination. This filter helps protect against the rist of cross-contamination and reduces maintenance requirements of the pipette. The filter tips are suited for molecular biology, microbiology, cell culture and radioactive work.
These tips have more space between the sample and the filter than conventional filter tips so you do not need to worry about the sample touching the filter whatever the pipetting technique or type of liquid.
Benefits:
  • Pipette foaming liquids like buffers and proteins
  • Pipette solvents
  • Reverse pipetting
  • Multiple dispensing functions of electronic pipettes
其他說明
Browse all pipette tips
從最近期的版本中選擇一個:
分析證明 (COA)