重要文件
Z740127
BRAND® pipette tips, racked, TipStack
volume 50-1000 μL, BIO-CERT®
登入查看組織和合約定價
About This Item
分類程式碼代碼:
41121600
暫時無法取得訂價和供貨情況
材料
colorless
colorless polypropylene tip
polypropylene
無菌
sterile; autoclaved
特點
RNase and DNase free
endotoxin free
BIO-CERT®
包裝
pack of 960 ea (10 racks of 96 + 2 boxes)
製造商/商標名
BRAND 732272
最大體積
1 mL
容積
50-1000 μL
尋找類似的產品? 前往 產品比較指南
一般說明
TipStack, non-sterile and sterile, BIO-CERT®
Pipette tips and filter tips are manufactured by BRAND in a cleanroom under the most modern production conditions, and are automatically rack packed and packaged to ensure that the tips are of consistently high quality.
A tip tower containing 5 filled tip-trays and a TipBox constitute the new, space-saving refill system for 20 μL, 200 μL and 1000 μL tips. Tightly sealing spacers prevent the tips from getting stuck together, and ensure them to be free from DNA, RNases, endotoxins and ATP.
The sterile TipStacks (BIO-CERT® quality) are supplied with a transfer aid for contamination-free use in a previously sterilized TipBox. Each packaging unit contains 2 TipStacks.
  • All components are recyclable
  • Reduced amount of waste
  • Sterilizable and reusable TipBox
  • High purity of the pipette tips and filter tips
  • Space-saving design
其他說明
Browse all pipette tips
法律資訊
BIO-CERT is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
從最近期的版本中選擇一個:
分析證明 (COA)