Key Documents
EP5452727007
Eppendorf® Rotor F-55-16-5-PCR
incl. rotor lid
登入查看組織和合約定價
About This Item
分類程式碼代碼:
41103909
NACRES:
NB.82
特點
to hold 2 x PCR strips
incl. rotor lid
包裝
pack of 1 ea
製造商/商標名
Eppendorf® 5452727007
技術
PCR: suitable
centrifugation: suitable
相容性
for use with centrifuge models MiniSpin/MiniSpin Plus
法律資訊
Eppendorf is a registered trademark of Eppendorf AG