重要文件
EP4720000011
Eppendorf® Varipette 4720
with coninuous volume adjustment 1-10 mL
登入查看組織和合約定價
About This Item
分類程式碼代碼:
41121500
NACRES:
NB.01
暫時無法取得訂價和供貨情況
特點
with coninuous volume adjustment 1-10 mL
製造商/商標名
4720000011
Eppendorf®
一般說明
Handle challenging liquid handling tasks with ease! The Varipette 4720 is a large volume, positive displacement pipette that is optimized for lab work with viscous or volatile liquids. Ideal for safely aspirating liquids from large bottles or tall, narrow vessels. Super easy, one-handed operation significantly speeds up pipetting tasks – simply press down the Varipettor operating lever to aspirate liquid from the reservoir; press the lever again and the liquid will be dispensed into your sample vessel.
特點和優勢
  • Mechanical positive displacement pipette designed for precise pipetting of liquids with high vapor pressure or viscosity
  • Volume range: 1 mL to 10 mL, adjustable in 10 µL increments for accurate volume setting via 4-digit volume display
  • Fast pipetting and convenient operation due to unique one-hand operation.
  • Liquid automatically aspirates and dispenses by simply pressing the operating lever
  • Eppendorf Varitips P is a syringe-style, positive displacement tip that allows for accurate pipetting of viscous solutions or liquids with high-vapor pressure
  • The Eppendorf Varitips S system, consisting of the dispensing part and Varitips, is optimized for aspirating liquids from large bottles or tall, narrow vessels
  • A valve for use with the Varitips S system ensures drip-free dispensing of volatile solutions
  • The graduation on Varitips ensures accurate dispensing of non-aqueous liquids
法律資訊
Eppendorf is a registered trademark of Eppendorf AG
從最近期的版本中選擇一個:
分析證明 (COA)