Key Documents
EP022654101
Lid for Eppendorf® FA-45-24-11-HS Rotor
aerosol-tight, FA-45-24-11-HS, 5430/5430R, aluminum, PTFE-coated
登入查看組織和合約定價
About This Item
分類程式碼代碼:
41103913
材料
PTFE-coated
aluminum
特點
aerosol-tight
包裝
pack of 1 ea
製造商/商標名
Eppendorf® 022654101
相容性
5430/5430R
FA-45-24-11-HS
一般說明
Rotor lid for FA-45-24-11-HS, aerosol-tight, aluminum, PTFE coated, for Centrifuge 5430/5430R
法律資訊
Eppendorf is a registered trademark of Eppendorf AG