重要文件
EP022260596
Accessories for Eppendorf® Pipettes & Repeater
wall mount, for Eppendorf Respeater Plus Pipete
登入查看組織和合約定價
About This Item
分類程式碼代碼:
41122801
暫時無法取得訂價和供貨情況
材料
plastic
包裝
pack of 1 ea
製造商/商標名
Eppendorf® 022260596
相關類別
一般說明
In addition to the pipette and Multipette® stands, a practical holder is available for wall mounting. This ensures that the instrument is correctly stored, always within reach, and does not take up valuable work space.
法律資訊
Eppendorf is a registered trademark of Eppendorf AG
Multipette is a registered trademark of Eppendorf AG
Repeater is a trademark of Brinkmann Instruments, Inc.
從最近期的版本中選擇一個:
分析證明 (COA)