Key Documents
BMSIPD96004PC
Sigma-Aldrich
BeadBlaster 96 Vial With Cap
polycarbonate, capacity 4 mL, 3/8” SS grinding ball
登入查看組織和合約定價
About This Item
分類程式碼代碼:
41100000
NACRES:
NA.82
材料
polycarbonate
製造商/商標名
Benchmark Scientific IPD9600-4PC
包裝
pack of 240 ×
容量
4 mL
相容性
3/8” SS grinding ball
一般說明
4ml polycarbonate vial with attached cap, for use with 3/8” stainless steel grinding ball in the BeadBlaster 96 homogenizer.
法律資訊
BeadBlaster is a trademark of Benchmark Scientific, Inc.