重要文件
YY2229353
Millipore
PTFE O-ring 293-Disc Filter Holder
登入查看組織和合約定價
About This Item
分類程式碼代碼:
41104909
eCl@ss:
32031613
暫時無法取得訂價和供貨情況
材料
PTFE
品質等級
相容性
for use with 293-mm Stainless Steel Sanitary Filter Holder
一般說明
The PTFE O-ring 293-Disc Filter Holder is intended for use with the 293 mm stainless steel sanitary filter holder - product number YY3029316 - an ideal solutions for sterilizing liquids or gases by pressure filtration.