167526
1.67526
1 of 10
SAFC
1.37009
氯化钾
Supelco
1.67529
Dip Tube(PVDF)for Sulfuric acid in 25 l fassett
1.04936
1.67525
Dip Tube (PE) for acids and bases in 25L canisters
901748
Dip tube
SYNF260810M
Synthware™ fritted gas dispersion tube
1.67521
Dip tube (PVDF) for sulfuric acid in 200 L PE-drums
C0100000
咖啡因
1.04165
1.04933
輸入產品批次/批號來搜索 分析證明 (COA)。在產品’s標籤上找到批次和批號,寫有 ‘Lot’或‘Batch’.。
產品編號
批次/批編號