重要文件
1.18344
Supelco
Lamp
登入查看組織和合約定價
About This Item
分類程式碼代碼:
41116116
NACRES:
SE.11
暫時無法取得訂價和供貨情況
一般說明
The lamp in the Turbiquant 1500 IR can be replaced if necessary. Only use the lamp that is intended for use with your instrument. For other turbidimeters please choose the respective lamp.
儲存和穩定性
Store at +5°C to +30°C.
法律資訊
TURBIQUANT is a trademark of Merck KGaA, Darmstadt, Germany