重要文件
Z609668
Bohlender screw caps
GL 25 thread
登入查看組織和合約定價
About This Item
分類程式碼代碼:
41121800
NACRES:
NB.17
暫時無法取得訂價和供貨情況
製造商/商標名
Bohlender
存貨情形
not available in USA and Canada
接頭
GL 25 thread
尋找類似的產品? 前往 產品比較指南
相關類別
一般說明
Made of PPS these screw caps with virgin PTFE membrane gaskets are resistant to high mechanical stresses - even at temperatures upto 250 °C. The gasket consists of PTFE membrane with a multidirectional orientated fibre pattern. Medium will only be exposed to PTFE.
法律資訊
Bohlender is a trademark of Bohlender GmbH
從最近期的版本中選擇一個:
分析證明 (COA)