重要文件
Z564001
Büchi® Rotavapor® RII evaporator with jack and water bath
joint: ST/NS 29/32, plastic-coated glass, condenser, diagonal, AC/DC input 230 - 240 V AC
登入查看組織和合約定價
About This Item
分類程式碼代碼:
41100000
暫時無法取得訂價和供貨情況
材料
plastic-coated glass
接合處
joint: ST/NS29/32
特點
CE compliant
CSA approved
condenser diagonal
製造商/商標名
Büchi 2411A1
尋找類似的產品? 前往 產品比較指南
一般說明
The Rotavapor RII fullfills exactly the fundamental needs of classical laboratory applications in teaching and research, practically and dependably, without compromising quality or safety. Available in three different glassware assemblies, all supplied with a standard 24/40 joint (US) or 29/32 (Europe) on the evaporator flask and a 35/20 ball joint on the receiving flask, and a choice of either standard glass or plastic-coated glass.
Model R II features:
  • Continuous vapor duct and a PTFE composite vacuum seal for long service life and effective sealing
  • Small footprint and robust design in powder-coated aluminum
  • Manual quick-action jack
  • Heating bath for temperatures of 0 to 95°C, quick-release electrical connection for easy removal
  • Process samples from 50 mL to 4 L
Evaporator with diagonal condenser assembly
For simple distillation of solvents and general use. Tap-water-cooled. Inlet feed tube for continuous feed of volumes exceeding flask capacity.
Evaporator with vertical condenser assembly
For standard distillations and for high-boiling solvents (>100°C). Tap-water-cooled. Inlet feed tube for continuous feed of volumes exceeding flask capacity.
Evaporator with cold-trap condenser assembly
Two-piece cold-trap condenser style is ideal for low-boiling solvents. Uses mechanical refrigeration or dry ice and acetone.
法律資訊
Büchi is a registered trademark of Büchi Labortechnik AG
Rotavapor is a registered trademark of Büchi Labortechnik AG
從最近期的版本中選擇一個:
分析證明 (COA)