重要文件
Z118982
Sigma-Aldrich
Scienceware® Frigimat® dry ice makers
USA connectors
登入查看組織和合約定價
About This Item
分類程式碼代碼:
41103000
暫時無法取得訂價和供貨情況
材料
aluminum ice chamber
製造商/商標名
Bel-Art
存貨情形
not available in EU
尋找類似的產品? 前往 產品比較指南
一般說明
A 6ft nickel plated copper connecting tube with USA standard CGA 320 inlet for attachment to CO2 cylinder is included.
Produces solid 1 lb cylindrical block 8.5 × 11 cm of dry ice from a cylinder of liquid CO2. One CO2 cylinder makes 8 to 9 blocks of dry ice. Epoxy-coated steel frame, brass valve, aluminum dry ice chamber.
法律資訊
Frigimat is a registered trademark of SP Scienceware, a division of SP
Scienceware is a registered trademark of SP Scienceware, a division of SP
從最近期的版本中選擇一個:
分析證明 (COA)