跳轉至內容
Merck
全部照片(1)

重要文件

VBP00038

Sigma-Aldrich

2-Chloro-8-cyclopentyl-5-methyl-8H-pyrido[2,3-d]pyrimidin-7-one

AldrichCPR

登入查看組織和合約定價


About This Item

經驗公式(希爾表示法):
C13H14ClN3O
分子量::
263.72
MDL號碼:
分類程式碼代碼:
12352200
PubChem物質ID:
暫時無法取得訂價和供貨情況

形狀

solid

SMILES 字串

CC(C1=CN=C(Cl)N=C1N2C3CCCC3)=CC2=O

InChI

1S/C13H14ClN3O/c1-8-6-11(18)17(9-4-2-3-5-9)12-10(8)7-15-13(14)16-12/h6-7,9H,2-5H2,1H3

InChI 密鑰

BSKNQSYIDZUXQT-UHFFFAOYSA-N

其他說明

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

象形圖

Exclamation mark

訊號詞

Warning

危險聲明

危險分類

Acute Tox. 4 Oral

儲存類別代碼

11 - Combustible Solids

水污染物質分類(WGK)

WGK 3

閃點(°F)

Not applicable

閃點(°C)

Not applicable


從最近期的版本中選擇一個:

分析證明 (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our 文件 section.

如果您需要協助,請聯絡 客戶支援

已經擁有該產品?

您可以在文件庫中找到最近購買的產品相關文件。

存取文件庫

Questions

Reviews

No rating value

Active Filters

我們的科學家團隊在所有研究領域都有豐富的經驗,包括生命科學、材料科學、化學合成、色譜、分析等.

聯絡技術服務