重要文件
TEM-LC300-AU-100
Sigma-Aldrich
Lacey Carbon Supported Gold Grid
size 300 mesh, hole size 100 μm , Average, box of 25
登入查看組織和合約定價
About This Item
分類程式碼代碼:
41111761
NACRES:
MA.24
暫時無法取得訂價和供貨情況
包裝
box of 25
孔尺寸
100 μm , Average
尺寸
300 mesh
尋找類似的產品? 前往 產品比較指南
一般說明
Lacey Carbon Supported Grid: Gold Grid, 300 mesh. Average hole size is 100 micron. This carbon coated film on a broken pattern consists of woven-mesh-like openings of different sizes and shapes. This type of pattern provides support but does not interfere when observing specimen sections.
從最近期的版本中選擇一個:
分析證明 (COA)