跳轉至內容
Merck
全部照片(1)

重要文件

L383384

4-[(E)-({[(2-CHLOROBENZOYL)AMINO]ACETYL}HYDRAZONO)METHYL]-2-METHOXYPHENYL 2-CHLOROBENZOATE

AldrichCPR

登入查看組織和合約定價


About This Item

線性公式:
C24H19Cl2N3O5
CAS號碼:
分子量::
500.342
MDL號碼:
暫時無法取得訂價和供貨情況

其他說明

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of rare and unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH'S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT AS-IS AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

從最近期的版本中選擇一個:

分析證明 (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our 文件 section.

如果您需要協助,請聯絡 客戶支援

已經擁有該產品?

您可以在文件庫中找到最近購買的產品相關文件。

存取文件庫

我們的科學家團隊在所有研究領域都有豐富的經驗,包括生命科學、材料科學、化學合成、色譜、分析等.

聯絡技術服務