重要文件
CLS70795N
Pyrex® serological pipettes, disposable, non-sterile, unplugged
volume 5.0 mL
同義詞:
pipet
登入查看組織和合約定價
About This Item
分類程式碼代碼:
41121500
NACRES:
NB.04
暫時無法取得訂價和供貨情況
材料
borosilicate glass
無菌
non-sterile
特點
DNase free: no
RNase free: no
graduations
subdivision
包裝
pack of 240 ea
case of 960 ea
製造商/商標名
Corning 7079-5N
體積測量範圍
5.0 mL
容量
5.0 mL
吸頭尺寸
, Narrow
尋找類似的產品? 前往 產品比較指南
相關類別
特點和優勢
  • Disposable PYREX® glass pipettes are calibrated "to deliver" (TD) with blow-out
  • Long, slender, tapered tips of pipettes make pipetting faster and easier
  • All pipettes have negative graduations and fire polished tips for burr-free, even flow rates
  • Nonsterile, unplugged and bulk packed.
法律資訊
PYREX is a registered trademark of Corning, Inc.
從最近期的版本中選擇一個:
分析證明 (COA)