重要文件
CLS5400125
Pyrex® iodine determination flask, with Pyrex® ST stopper
capacity 125 mL
同義詞:
flask erlenmeyer, flasks erlenmeyer, glass flasks, Corning® 5400
登入查看組織和合約定價
About This Item
分類程式碼代碼:
41121804
NACRES:
NB.43
暫時無法取得訂價和供貨情況
材料
borosilicate glass
wide-mouth
接合處
joint ST/NS stopper22
無菌
non-sterile
特點
closure type ST stopper
包裝
case of 12
製造商/商標名
Corning 5400-125
容量
125 mL
尋找類似的產品? 前往 產品比較指南
一般說明
Flasks are blown in specillay designed molds, thereby insuring uniformity and proper contour. The stopper projects above the liquid seal trough to facilitate removal.
法律資訊
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
從最近期的版本中選擇一個:
分析證明 (COA)