Merck
全部照片(3)
文件
743992
Sigma-Aldrich
Glass microreactor LTF MR Lab VS
with C-profile
登入查看組織和合約定價
About This Item
分類程式碼代碼:
12162002
NACRES:
NA.22
品質等級
一般說明
  • Volume 1.1 ml
  • Delay time
  • For mixture-intensive substances
  • Max pressure: 15 bar
  • With aluminium connecting bar