Skip to Content
Merck
All Photos(2)

Key Documents

MAK203

Sigma-Aldrich

DPP4 INHIBITOR SCREENING KIT

Sufficient for 100 Fluorometric tests

Synonym(s):

Adenosine Deaminase Complexing Protein-2 Inhibitor Screening Kit, CD26 Inhibitor Screening Kit, Dipeptidyl-Peptidase 4 Inhibitor Screening Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.25
Pricing and availability is not currently available.

application(s)

pharmaceutical

detection method

fluorometric

relevant disease(s)

endocrinological disorders, diabetes

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... DPP4(1803)
mouse ... DPP4(13482)
rat ... DPP4(25253)

General description

Dipeptidyl-peptidase 4 (DPP4; CD26; adenosine deaminase complexing protein-2) is a membrane glycoprotein with serine exopeptidase activity that cleaves X-proline dipeptides from the N-terminus of polypeptides. Inhibitors of DPP4 inhibit the degradation of glucose-dependent insulinotropic polypeptide and glucagon-like peptide-1 by DPP4 and have emerged as oral anti-diabetic agents.

Features and Benefits

Compatible with high-throughput handling systems.

Suitability

Suitable for the screening of inhibitors of DPP4.

Principle

The DPP4 Inhibitor Screening Kit is a simple assay suitable for high-throughput screening of potential DPP4 inhibitors. DPP4 activity is measured by cleaving the substrate to yield a fluorescent product (λex = 360/λem = 460 nm), proportional to the enzymatic activity present. The effectiveness of the test inhibitors may be compared with the DPP4 inhibitor (sitagliptin) provided as a control.

Kit Components Only

Product No.
Description

  • DPP4 Assay Buffer

  • DPP4 Substrate

  • DPP4 Enzyme

  • DPP4 Inhibitor, Sitagliptin

Storage Class Code

10 - Combustible liquids

Flash Point(F)

188.6 °F - closed cup

Flash Point(C)

87 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cleni Palma-Albino et al.
Antioxidants (Basel, Switzerland), 10(11) (2021-11-28)
Multifunctional peptides, capable of acting on different body systems through multiple mechanisms of action, offer many advantages over monofunctional peptides, including lower adverse side effects and costs. Erythrina edulis (pajuro) is a legume with a large number of high-quality proteins
Bingli Zhao et al.
Bioorganic chemistry, 99, 103772-103772 (2020-03-29)
Two key enzymes like α-glucosidase and dipeptidyl peptidase-4 (DPP4) play an important role in the development of Type 2 diabetes (T2D), and inhibiting them is a common treatment strategy for T2D. In this study, an ultrafiltration- centrifugation approach was developed
Paulina Kęska et al.
Nutrients, 14(4) (2022-02-27)
The ability of peptides from an aqueous and salt-soluble protein extract of dry-cured pork loins to inhibit the action of dipeptidyl peptidase IV was determined. This activity was assessed at different times of the production process, i.e., 28, 90, 180
S Wupper et al.
Journal of physiology and pharmacology : an official journal of the Polish Physiological Society, 71(3) (2020-09-30)
Gamma-cyclodextrin (γCD) is a cyclic oligosaccharide consisting of eight α-(1,4)-linked glucopyranose subunits, which is often used in the food and pharmaceutical industries. However, little is known regarding the metabolic activity of "empty" γCD per se. Therefore, in the present study

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service