Skip to Content
Merck
All Photos(1)

Key Documents

159328

Sigma-Aldrich

1,4-Dichloro-2-butene (cis+trans)

technical grade, 90%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCH2CH=CHCH2Cl
CAS Number:
Molecular Weight:
125.00
Beilstein:
1361446
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
Pricing and availability is not currently available.

grade

technical grade

Assay

90%

functional group

chloro

storage temp.

2-8°C

SMILES string

C\C(Cl)=C/CCl

InChI

1S/C4H6Cl2/c1-4(6)2-3-5/h2H,3H2,1H3/b4-2+

InChI key

WLIADPFXSACYLS-DUXPYHPUSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

127.4 °F - closed cup

Flash Point(C)

53 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Phadtare et al.
Nucleic acids symposium series, (18)(18), 25-28 (1987-01-01)
Reaction of adenine (1a) or cytosine (1b) with excess 1,4-dichloro-2-butyne catalyzed by K2CO3 in (CH3)2SO gave the 4-chloro-2-butynyl derivatives 2a and 2b. The latter were converted to the 4-hydroxy-2-butynyl compounds 3a and 3b by refluxing in 0.1 M HCl. Isomerization
[Experimental data on a hygienic standard for 1,4-dichlorobutene-2 in the air of a work area].
M S Gizhlarian et al.
Gigiena truda i professional'nye zabolevaniia, (4)(4), 49-50 (1985-04-01)
L S Mullin et al.
Drug and chemical toxicology, 23(3), 403-417 (2000-08-26)
This study was conducted to elucidate the time- and dose-response relationships of long-term, low-level 1,4-dichlorobutene-2 (DCB) inhalation exposure to nasal tumor induction in rats. Male Crl:CD BR rats were exposed 6 hours per day, 5 days week to 0, 0.1
L S Mullin et al.
Drug and chemical toxicology, 25(2), 227-230 (2002-05-25)
Rats were exposed by inhalation to either 0.5 ppm 1,4-dichlorobutene-2 (DCB) for two years or to 5.0 ppm for seven months, 2.5 ppm for five months, and no further exposure for 12 months prior to sacrifice. Malignant and non-malignant tumors
Teratogenic evaluation of 1,4-dichlorobutene-2 in the rat following inhalation exposure.
G L Kennedy et al.
Toxicology and applied pharmacology, 64(1), 125-130 (1982-06-15)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service