Skip to Content
Merck
All Photos(1)

Key Documents

662410

Sigma-Aldrich

Lumidot CdS

core-type quantum dots, 5 mg/mL in toluene

Synonym(s):

Cadmium monosulfide, Cadmium sulfide yellow

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CdS
CAS Number:
Molecular Weight:
144.48
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:

form

dispersion

concentration

5 mg/mL in toluene

particle size

1.8-2.3 nm

density

0.865 g/mL at 25 °C

λmax

370-390 nm

fluorescence

λem 390-410 nm FWHM 18 nm

storage temp.

2-8°C

InChI

1S/Cd.S

InChI key

CJOBVZJTOIVNNF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Organically stabilized cadmium sulfide quantum dots in toluene

Analysis Note

Particle size was determined by TEM according to the procedure in Yu, W. W., et al., Chem. Mater., 2003, 15, 2854.
Quantum yield was measured following the procedure of Qu, L., Peng, X., J. Am. Chem. Soc., 2002, 124-2049.

Other Notes

Lifetime one year if stored properly

Legal Information

Product of Nanoco Technologies sold under US and worldwide patents and patents pending.
Lumidot is a trademark of Sigma-Aldrich Co. LLC

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Carc. 1B - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

39.2 °F

Flash Point(C)

4 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mark P Hendricks et al.
ACS nano, 6(11), 10054-10062 (2012-10-10)
We describe the synthesis of cadmium bis(diphenyldithiophosphinate) (Cd(S(2)PPh(2))(2)) from secondary phosphine sulfides and its conversion to cadmium sulfide nanocrystals. Heating Cd(S(2)PPh(2))(2) and cadmium tetradecanoate (≥4 equiv) to 240 °C results in complete conversion of Cd(S(2)PPh(2))(2) to cadmium sulfide nanocrystals with
Huan-Wei Tseng et al.
Journal of the American Chemical Society, 135(9), 3383-3386 (2013-02-15)
We describe the charge transfer interactions between photoexcited CdS nanorods and mononuclear water oxidation catalysts derived from the [Ru(bpy)(tpy)Cl](+) parent structure. Upon excitation, hole transfer from CdS oxidizes the catalyst (Ru(2+) → Ru(3+)) on a 100 ps to 1 ns
Massimo Rippa et al.
Nanoscale, 5(1), 331-336 (2012-11-21)
In this paper two-dimensional (2D) photonic Thue-Morse quasicrystals (ThMo-PQCs) in active CdSe/CdS nanorod (NR) doped polymer nanocomposites are proposed and experimentally demonstrated. Active PQCs and undoped lattices have been prepared in a one-step fabrication process by an electron beam lithography
Jianjun Tian et al.
Nanoscale, 5(3), 936-943 (2012-11-21)
Photoelectrode made of nanocable structure of ZnO nanorods (NR) coated with TiO(2) nanosheets (NSs) was investigated for CdS/CdSe quantum dot co-sensitized solar cells. ZnO NRs prepared solution reaction at 60 °C served as the backbone for direct electron transport in
Chengzhen Wei et al.
Chemphyschem : a European journal of chemical physics and physical chemistry, 14(3), 591-596 (2013-01-09)
In this study, we report the synthesis of monodispersive solid and hollow CdS spheres with structure-dependent photocatalytic abilities for dye photodegradation. The monodispersive CdS nanospheres were constructed with the assistance of the soulcarboxymthyi chitosan biopolymer under hydrothermal conditions. The solid

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service