Merck
  • The importance of nanocrystal precursor conversion kinetics: mechanism of the reaction between cadmium carboxylate and cadmium bis(diphenyldithiophosphinate).
The importance of nanocrystal precursor conversion kinetics: mechanism of the reaction between cadmium carboxylate and cadmium bis(diphenyldithiophosphinate).
ACS nano (2012-10-10)
Mark P Hendricks, Brandi M Cossairt, Jonathan S Owen
ABSTRACT
We describe the synthesis of cadmium bis(diphenyldithiophosphinate) (Cd(S(2)PPh(2))(2)) from secondary phosphine sulfides and its conversion to cadmium sulfide nanocrystals. Heating Cd(S(2)PPh(2))(2) and cadmium tetradecanoate (≥4 equiv) to 240 °C results in complete conversion of Cd(S(2)PPh(2))(2) to cadmium sulfide nanocrystals with tetradecanoate surface termination. The nanocrystals have a narrow size distribution (d = 3.8-4.1 nm, σ < 10%) that is evident from the line width of the lowest energy absorption feature (λ = 412-422 nm, fwhm = 0.17 eV) and display bright photoluminescence (PLQY(band edge+trap) = 36%). Interestingly, the final diameter is insensitive to the reaction conditions, including the total concentration of precursors and initial cadmium to sulfur ratio. Monitoring the reaction with (31)P NMR, UV-visible, and infrared absorption spectroscopies shows that the production of cadmium diphenylphosphinate (Cd(O(2)PPh(2))(2)) and tetradecanoic anhydride co-products is coupled with the formation of cadmium sulfide. From these measurements we propose a balanced chemical equation for the conversion reaction and use it to optimize a synthesis that affords CdS nanocrystals in quantitative yield. In light of these results we discuss the importance of well-defined precursor reactivity to reproducible conversion kinetics and the synthesis of nanocrystals with unambiguous chemical composition.
Sigma-Aldrich
Lumidot CdS, core-type quantum dots, 5 mg/mL in toluene
Sigma-Aldrich
Cadmium sulfide, powder, 99.995% trace metals basis