Merck
  • Charge transfer dynamics between photoexcited CdS nanorods and mononuclear Ru water-oxidation catalysts.
Charge transfer dynamics between photoexcited CdS nanorods and mononuclear Ru water-oxidation catalysts.
Journal of the American Chemical Society (2013-02-15)
Huan-Wei Tseng, Molly B Wilker, Niels H Damrauer, Gordana Dukovic
ABSTRACT
We describe the charge transfer interactions between photoexcited CdS nanorods and mononuclear water oxidation catalysts derived from the [Ru(bpy)(tpy)Cl](+) parent structure. Upon excitation, hole transfer from CdS oxidizes the catalyst (Ru(2+) → Ru(3+)) on a 100 ps to 1 ns timescale. This is followed by 10-100 ns electron transfer (ET) that reduces the Ru(3+) center. The relatively slow ET dynamics may provide opportunities for the accumulation of multiple holes at the catalyst, which is necessary for water oxidation.
Sigma-Aldrich
Lumidot CdS, core-type quantum dots, 5 mg/mL in toluene
Sigma-Aldrich
Cadmium sulfide, powder, 99.995% trace metals basis