主要文件
57630
7725 型进样器定量环
volume 1 mL
登录查看公司和协议定价
选择尺寸
1 EA
$110.00
$110.00
预计发货时间2025年5月22日详情
获取大包装报价
选择尺寸
变更视图
1 EA
$110.00
About This Item
UNSPSC代码:
41115854
$110.00
预计发货时间2025年5月22日详情
获取大包装报价
物料
stainless steel sample loop and fittings
制造商/商品名称
Rheodyne 7755-027
体积
1 mL
相容性
for use with Rheodyne 7725 and 7725i injector
正在寻找类似产品? 访问 产品对比指南
一般描述
Sample loop for model 7725 injector is designed for advanced fluid-handling solutions for a wide range of analytical instrumentation and in vitro diagnostic systems. It can be used in high performance liquid chromatographic systems and is quite accurate and precise in injecting 1μL-5 mL concentration of samples.
推荐产品
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
历史批次信息供参考:
分析证书(COA)
访问文档库
商品
Correct common HPLC issues to restore column performance, addressing peak problems, retention time, noise, and resolution with chromatograms.
识别、纠正常见HPLC问题,恢复色谱柱性能。通过色谱图示例,获取解决峰相关问题(峰分裂、峰拖尾、峰前延等)、保留时间、吹扫噪声及分离度下降问题排查帮助。