504343
1 of 10
Supelco
504394
Empty glass SPE Tube with PTFE frit, 20 μm porosity
57242
Empty SPE Tube (no frits)
Sigma-Aldrich
591742
苯酚-2,4,6-d3,OD
Millipore
77730
革兰氏染色试剂盒
P4557
苯酚 溶液
504335
SPE 小柱连接头
57177
带 PE 筛板的聚丙烯固相萃取空管,20μm 孔隙度
57020-U
802913
(24R),24,25-Dihydroxyvitamin D3-26,26,26,27,27,27-d6 solution
57026
solid PTFE
pk of 24
solid phase extraction (SPE): suitable
food and beverages
for use with 6mL glass SPE tube
从最新的版本中选择一种:
抱歉,我们目前尚未线上提供该产品的COA。