Merck
2631005
Supelco
进样口衬管,分流式(填充玻璃棉),用于 AutoSystem
pkg of 5 ea
登录查看公司和协议定价
About This Item
分類程式碼代碼:
12000000
包裝
pkg of 5 ea
長度 × 外徑 × 內徑
92 mm × 6.35 mm × 4.0 mm
正在寻找类似产品? 访问 产品对比指南
一般說明
Deactivated (silanized) glass inlet liner for capillary injection ports.
  • Fits PerkinElmer (AutoSystem)
Choose from one of the most recent versions:
分析证书(COA)
Lot/Batch Number
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact 客户支持