主要文件
Z743565
Petri-Seal adhesive sealing film
red, L × W 12.5 in. × 32.9 in.
登录查看公司和协议定价
选择尺寸
1 EA
$56.60
$56.60
预计发货时间2025年6月18日详情
选择尺寸
变更视图
1 EA
$56.60
About This Item
UNSPSC代码:
41122410
NACRES:
NB.35
$56.60
预计发货时间2025年6月18日详情
包装
pack of 1 ea
制造商/商品名称
Diversified Biotech PSEAL-108R
长度 × 宽度
12.5 in. × 32.9 in.
颜色
red
正在寻找类似产品? 访问 产品对比指南
一般描述
A sealing tape designed specifically for petri dishes and multiwell plates.
Conforms to the irregular perimeter to seal cover to plate, reduce evaporation, and prevent contamination. Waterproof, chemically resistant, and suitable for continuous use from −40 to +77°C. Heavy 5-mil (0.125 mm) thickness resists abrasion and puncture.
法律信息
Petri-Seal is a trademark of C.B.S. Scientific Co.
历史批次信息供参考:
分析证书(COA)