主要文件
Z741972
Cameo Filtration Membrane
Nylon membrane, pore size 0.45 μm, diam. 47 mm, pkg of 100 ea
登录查看公司和协议定价
选择尺寸
100 EA
$312.00
$312.00
请联系客服了解存货情况
选择尺寸
变更视图
100 EA
$312.00
About This Item
UNSPSC代码:
41104900
NACRES:
NB.24
$312.00
请联系客服了解存货情况
物料
Nylon membrane
无菌性
non-sterile
包装
pkg of 100 ea
制造商/商品名称
GVS Life Sciences 1213776
直径
47 mm
孔径
0.45 μm
正在寻找类似产品? 访问 产品对比指南
一般描述
Designed to speed up and increase sample volume throughput while reducing back pressure, the hold-up volume is slightly lower due to improved flow channel design and reduced spacing between the supports. The design improvements allow an increase in operating pressure the syringe filters can withstand.
Applications: Analysis of Biological and Organic Fluids, Analytical Sample Prep, HPLC, Composite assays, Content uniformity, Dissolution testing, Environmental samples, Food Analysis
法律信息
Cameo is a trademark of GVS S.p.A.
历史批次信息供参考:
分析证书(COA)