主要文件
Z325368
BRAND® BLAUBRAND® bulb pipette, calibrated to deliver (TD, EX)
capacity 20 mL, two marks
登录查看公司和协议定价
About This Item
UNSPSC代码:
41121500
NACRES:
NB.01
产品Z325368监管中,请联系销售询价或购买 联系技术服务部门
物料
AR-Glas®
描述
partial delivery
特点
class AS
包装
pack of 6 ea
制造商/商品名称
BRAND 29734
体积测量范围
20 mL, accuracy: 0.03 mL
容量
20 mL
正在寻找类似产品? 访问 产品对比指南
一般描述
All BLAUBRAND bulb pipettes are supplied with a batch number and an accompanying batch certificate in the original packaging.
According to DIN EN ISO 648.
其他说明
BR29734 (India)
法律信息
AR-Glas is a registered trademark of Schott AG
BLAUBRAND is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
历史批次信息供参考:
分析证书(COA)
访问文档库
实验方案
本页面包含操作移液管和助吸器的详细信息。
This page contains detailed information on working with and handling pipettes and pipetting aids.