跳转至内容
Merck

T2199

Sigma-Aldrich

Triton X-100

别名:

4-(1,1,3,3-四甲基丁基)苯基-聚乙二醇, t-辛基苯氧基聚乙氧基乙醇, 聚乙二醇 辛基苯醚

登录查看公司和协议定价


About This Item

线性分子式:
t-Oct-C6H4-(OCH2CH2)xOH, x= 9-10
CAS号:
Beilstein:
2315025
MDL號碼:
分類程式碼代碼:
12352200
价格与库存信息目前不能提供

分子量

micellar avg mol wt 80,000
average mol wt 625

凝集数

100-155

CMC

0.2-0.9 mM (20-25°C)

轉變溫度

cloud point 65 °C
pour point ~7 °C

HLB

13.5

SMILES 字串

CC(C)(C)CC(C)(C)c1ccc(OCCOCCOCCOCCOCCOCCOCCO)cc1

InChI

1S/C28H50O8/c1-27(2,3)24-28(4,5)25-6-8-26(9-7-25)36-23-22-35-21-20-34-19-18-33-17-16-32-15-14-31-13-12-30-11-10-29/h6-9,29H,10-24H2,1-5H3

InChI 密鑰

HNLXNOZHXNSSPN-UHFFFAOYSA-N

正在寻找类似产品? 访问 产品对比指南

生化/生理作用

广泛使用的非离子型表面活性剂,用于在温和的非变性条件下回收膜组分。

法律資訊

Triton is a trademark of The Dow Chemical Company or an affiliated company of Dow

取代透過

产品编号
说明
价格

訊號詞

Danger

危險分類

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Irrit. 2

儲存類別代碼

10 - Combustible liquids

水污染物質分類(WGK)

WGK 3

閃點(°F)

483.8 °F - closed cup

閃點(°C)

251 °C - closed cup

個人防護裝備

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


历史批次信息供参考:

分析证书(COA)

Lot/Batch Number

没有发现合适的版本?

如果您需要特殊版本,可通过批号或批次号查找具体证书。

已有该产品?

在文件库中查找您最近购买产品的文档。

访问文档库

Friederike Pfeiffer et al.
Acta neuropathologica, 122(5), 601-614 (2011-10-11)
In experimental autoimmune encephalomyelitis (EAE), an animal model for multiple sclerosis (MS), loss of the blood-brain barrier (BBB) tight junction (TJ) protein claudin-3 correlates with immune cell infiltration into the CNS and BBB leakiness. Here we show that sealing BBB
Adam A Strömstedt et al.
Antimicrobial agents and chemotherapy, 53(2), 593-602 (2008-11-26)
Methods for increasing the proteolytic stability of EFK17 (EFKRIVQRIKDFLRNLV), a new peptide sequence with antimicrobial properties derived from LL-37, were evaluated. EFK17 was modified by four d-enantiomer or tryptophan (W) substitutions at known protease cleavage sites as well as by
Manuela Rinaldi et al.
Veterinary research, 42, 61-61 (2011-05-17)
The mucus layer in the gastrointestinal (GI) tract is considered to be the first line of defense to the external environment. Alteration in mucus components has been reported to occur during intestinal nematode infection in ruminants, but the role of
Andri K Riau et al.
Molecular vision, 15, 335-342 (2009-02-19)
Pterygium is an ocular surface disease of unknown etiology associated with epithelial and fibrovascular outgrowth from the conjunctiva onto the cornea. S100 proteins are calcium-activated signaling proteins that interact with other proteins to modulate biological functions such as cell migration
Lionel Rigottier-Gois et al.
PloS one, 6(12), e29023-e29023 (2011-12-24)
Spread of antibiotic resistance among bacteria responsible for nosocomial and community-acquired infections urges for novel therapeutic or prophylactic targets and for innovative pathogen-specific antibacterial compounds. Major challenges are posed by opportunistic pathogens belonging to the low GC% gram-positive bacteria. Among

我们的科学家团队拥有各种研究领域经验,包括生命科学、材料科学、化学合成、色谱、分析及许多其他领域.

联系技术服务部门