HS120316
$29.20
国内现货,预计发货时间2025年4月14日详情
1 of 10
HS15990A
Slide box
HS15991A
BR476000
BRAND®涂片盒
HS15991B
Sigma-Aldrich
Z374385
Scienceware ® 载玻片盒
BR475900
HS15991C
HS15990C
HS15994E
Premium slide box
HS15988H
TrueNorth® Microscope Slide Box
blue polycarbonate
Holds 25 x slides 76x26 mm
pack of 1 ea
Heathrow Scientific HS120316
141 × 88 × 35 mm
正在寻找类似产品? 访问 产品对比指南
样品处理
历史批次信息供参考:
It looks like we've run into a problem, but you can still download Certificates of Analysis from our 文件 section.
如需帮助,请联系 客户支持
在文件库中查找您最近购买产品的文档。
访问文档库
1 of 8
HS159890B
Storage rack for 50 and 100 place microscope slide boxes
HS15994F
HS15994G
Premium Slide box
HS15994A
Z742390
PELCO® Sliding Latch Microslide Storage Boxes