主要文件
EP5920748009
Adapter for Eppendorf® S-4x Universal Large Rotor
capacity, 7 x 50 mL conical tubes, pack of 2
登录查看公司和协议定价
About This Item
UNSPSC代码:
41103911
NACRES:
NB.82
特点
capacity 7 x 50 mL conical tubes
Holds 50 mL tubes
包装
pack of 2
制造商/商品名称
Eppendorf® 5920748009 (Global)
技术
centrifugation: suitable
相容性
for use with Universal-Large bucket
正在寻找类似产品? 访问 产品对比指南
一般描述
Adapters for various sizes of tubes and bottles for use with Eppendorf S-4x Universal large rotor.
法律信息
Eppendorf is a registered trademark of Eppendorf AG
历史批次信息供参考:
分析证书(COA)