主要文件
BMSIPD960035ZO
Sigma-Aldrich
BeadBlaster 96 Grinding Jars for Trace Metal Samples
(zirconium oxide), capacity 35 mL, pack of 2
登录查看公司和协议定价
选择尺寸
2 EACH
$8,140.00
$8,140.00
预计发货时间2025年6月25日详情
选择尺寸
变更视图
2 EACH
$8,140.00
About This Item
UNSPSC代码:
41100000
NACRES:
NA.82
$8,140.00
预计发货时间2025年6月25日详情
物料
(zirconium oxide)
包装
pack of 2
制造商/商品名称
Benchmark Scientific IPD9600-35ZO
容量
35 mL
相容性
BeadBlaster 96
一般描述
The jars feature threaded closures for leakproof performance with both solids and liquids.
Zirconium oxide is a chemically inert ceramic material. Not as hard as and less expensive than tungsten carbide, it is used for softer samples where trace metals from stainless steel may be a concern. These jars are ideal for grinding biological samples, plants, bones, teeth, soils and sludges. Jars are offered in a 35ml volume.
法律信息
BeadBlaster is a trademark of Benchmark Scientific, Inc.
历史批次信息供参考:
分析证书(COA)